Learn More
Raybiotech Inc Mouse Galectin-1 ELISA Kit
Supplier:  Raybiotech Inc ELMGALECTIN11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 12 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes mouse Galectin-1.
Catalog No. 50-194-9993