Learn More
Abcam Mouse Galectin 1 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1195951X96TESTS
Encompass_Preferred
Mouse Galectin 1 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Galectin 1 in Mouse in Plasma, Cell culture supernatant, Serum, Cell Lysate samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2587590