Learn More
Biomatik Corporation Mouse Galectin 12 (GAL12) ELISA Kit
Supplier:  Biomatik Corporation EKU04317
Encompass_Preferred
Mouse Galectin 12 (GAL12) ELISA Kit for detecting GAL12 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91VD1. Gene ID: 56072. Target Synonyms: LGALS12; GRIP1; Lectin,Galactoside-Binding Soluble 12; Galectin-related inhibitor of proliferation. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5941