Learn More
Biomatik Corporation Mouse Galectin 9 (GAL9) CLIA Kit
Supplier:  Biomatik Corporation EKU10855
Encompass_Preferred
Mouse Galectin 9 (GAL9) CLIA Kit for detecting GAL9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O08573. Gene ID: 16859. Target Synonyms: LGALS9; Ecalectin; Lectin,Galactoside-Binding Soluble 9; Tumor antigen HOM-HD-21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8278