Learn More
Biomatik Corporation Mouse Gamma-Endorphin (gEP) ELISA Kit
Supplier:  Biomatik Corporation EKU04333
Encompass_Preferred
Mouse Gamma-Endorphin (gEP) ELISA Kit for detecting gEP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1000pg/ml. Sensitivity: 4.72pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01193. Gene ID: 18976. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5977