Learn More
Biomatik Corporation Mouse Gamma-Glutamyltransferase 1 (gGT1) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10549
Encompass_Preferred
Mouse Gamma-Glutamyltransferase 1 (gGT1) Wide-range ELISA Kit for detecting gGT1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60928. Gene ID: 14598. Target Synonyms: CD224; G-GT1; GTG; Gamma-glutamyltranspeptidase 1; Glutathione hydrolase 1; Leukotriene-C4 hydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8330