Learn More
Biomatik Corporation Mouse Gasdermin D (GSDMD) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11641
Encompass_Preferred
Mouse Gasdermin D (GSDMD) Mini sample ELISA Kit for detecting GSDMD in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D8T2. Gene ID: 69146. Target Synonyms: GSDMDC1; DF5L; Gasdermin Domain Containing 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8111