Learn More
Abcam Mouse GDF-15 Matched Antibody Pair Kit, 5 x 96 Tests
Catalog No. 502657471
Encompass_Preferred
Mouse GDF-15 Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Mouse GDF-15 by sandwich ELISA method.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7471 Supplier Abcam Supplier No. AB2201315X96TESTS
Mouse GDF-15 Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Mouse GDF-15 by sandwich ELISA method.
The product is subject to the following: Abcam Restricted Use Statement