Learn More
Sino Biological Recombinant Human, Mouse, Rat, Rabbit GDF-8/Myostatin Protein (hFc Tag)
Supplier:  Sino Biological 50441M01H50
Encompass_Preferred
A DNA sequence encoding the mature form of Human, Mouse, Rat, Rabbit MSTN (NP_034964.1) (Asp 268-Ser 376) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-224-7250