Learn More
Apexbio Technology LLC Direct Mouse Genotyping Kit, 500 rxns. Cas: MFCD: NA.
SDP
Catalog No. 501905570
Encompass_Preferred
The Direct Mouse Genotyping Kit for genotyping provides convenience in faster preparation and superior PCR amplification. Lysis buffer and balance buffer could digest mice tissues rapidly to release unbroken genomic DNA, and it could be used as PCR template directly without extraction from the mixed solution. By using our kit, you can substantially shorten your time for digestion. Furthermore, the 2PCR Master Mix (With Dye) will guarantee accurate amplification consequence. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5570 Supplier Apexbio Technology LLC Supplier No. K1025500
The Direct Mouse Genotyping Kit for genotyping provides convenience in faster preparation and superior PCR amplification. Lysis buffer and balance buffer could digest mice tissues rapidly to release unbroken genomic DNA, and it could be used as PCR template directly without extraction from the mixed solution. By using our kit, you can substantially shorten your time for digestion. Furthermore, the 2PCR Master Mix (With Dye) will guarantee accurate amplification consequence. Other sizes are available. Please inqury us for quote.