Learn More
Apexbio Technology LLC Direct Mouse Genotyping Kit Plus, 500 rxns
SDP
Catalog No. 501998269
Encompass_Preferred
The Direct Mouse Genotyping Kit plus provides convenience in faster preparation and superior PCR amplification. Lysis buffer and balance buffer could digest mice tissues rapidly to release unbroken genomic DNA, and it will be used as PCR template directly without extraction from the mixed solution. By using our kit, you can substantially shorten your time for digestion. Furthermore, the 2PCR Master Mix plus (With Dye) will guarantee accurate amplification consequence. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8269 Supplier Apexbio Technology LLC Supplier No. K1027500
The Direct Mouse Genotyping Kit plus provides convenience in faster preparation and superior PCR amplification. Lysis buffer and balance buffer could digest mice tissues rapidly to release unbroken genomic DNA, and it will be used as PCR template directly without extraction from the mixed solution. By using our kit, you can substantially shorten your time for digestion. Furthermore, the 2PCR Master Mix plus (With Dye) will guarantee accurate amplification consequence. Other sizes are also available. Please inqury us for quote.