Learn More
Sino Biological Anti-GFR Alpha-1/GFRA1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50171RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse GFRa1 / GFR alpha-1 (rM GFRA1; Catalog#50171-M08H; AAB86600.1; Met 1-Ser 425). GFRa1 specific IgG was purified by mouse GFRa1 affinity chromatography.
Catalog No. 50-255-3391