Learn More
Sino Biological Anti-GFR Alpha-3/GFRA3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106718T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse GFRA3, and purified by antigen affinity chromatography.
Catalog No. 50-241-0674