Learn More
Crystal Chem Inc Mouse Growth Hormone (GH) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80587
Encompass
The Mouse Growth Hormone ELISA is used to quantify levels of mouse growth hormone in serum using only a 25 µL sample volume. Highlights: Kit uses sample size of only 25 µL. Dynamic range of 0.15 - 9 ng/mL. High sensitivity of 0.04 ng/mL. Designed for mouse samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7753