Learn More
Sino Biological Anti-Growth Hormone Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50043RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Growth Hormone Receptor / GHR / GHBP (rM Growth Hormone Receptor / GHR / GHBP; Catalog#50043-M08H; NP_034414.2; Met 1-Gln 273). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3522