Learn More
Sino Biological Mouse GHR PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50043RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Growth Hormone Receptor / GHR / GHBP (rM Growth Hormone Receptor / GHR / GHBP; Catalog#50043-M08H; NP_034414.2; Met 1-Gln 273). Growth Hormone Receptor / GHR / GHBP specific IgG was purified by Mouse Growth Hormone Receptor / GHR / GHBP affinity chromatography.
Catalog No. 50-237-3212