Learn More
Biomatik Corporation Mouse Ghrelin (GHRL) ELISA Kit
Supplier:  Biomatik Corporation EKU10865
Encompass_Preferred
Mouse Ghrelin (GHRL) ELISA Kit for detecting GHRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EQX0. Gene ID: 58991. Target Synonyms: MTLRP; Growth Hormone-Releasing Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7661