Learn More
BIOHIPPO Mouse GHRL Microsample
SDP
Catalog No. 5000313956
Encompass
Mouse GHRL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GHRL in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 33 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GHRL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GHRL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GHRL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13956 Supplier BIOHIPPO Supplier No. BHE15210763
Mouse GHRL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GHRL in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 33 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GHRL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GHRL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GHRL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color