Learn More
American Research Products Inc Mouse glial cell line deriVed
Catalog No. 5000328842
Encompass
Mouse glial cell line-deriVed neurotrophic factor GDNF ELISA KIT from CUSABIO Detection Range 10 9 pg/ml - 8000 pg/ml Sensitivity 10 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28842 Supplier American Research Products Inc Supplier No. CSBE07341M
Mouse glial cell line-deriVed neurotrophic factor GDNF ELISA KIT from CUSABIO Detection Range 10 9 pg/ml - 8000 pg/ml Sensitivity 10 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich