Learn More
Biomatik Corporation Mouse Glial Cell Line Derived Neurotrophic Factor (GDNF) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11131
Encompass_Preferred
Mouse Glial Cell Line Derived Neurotrophic Factor (GDNF) Mini sample ELISA Kit for detecting GDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48540. Gene ID: 14573. Target Synonyms: ATF1; ATF2; HFB1-GDNF; Astrocyte-Derived Trophic Factor; Glial Derived Neurotrophic Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4823