Learn More
American Research Products Inc Mouse glial fibrillary acidic
Catalog No. 5000330235
Encompass
Mouse glial fibrillary acidic protein GFAP ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30235 Supplier American Research Products Inc Supplier No. CSBE08603M
Mouse glial fibrillary acidic protein GFAP ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich