Learn More
Sino Biological Mouse GLIS1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 103978T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse GLIS1, and purified by antigen affinity chromatography.
Catalog No. 50-237-1998