Learn More
Crystal Chem Inc Mouse GLP-2 ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 81514
Encompass
The Mouse GLP-2 ELISA is a highly sensitive assay that is used to quantify levels of mouse GLP-2 in serum and plasma. Highlights: Kits use a sample volume of only 25 µL. High Sensitivity of 0.41 ng/mL. Dynamic range of 0.41 - 100 ng/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7900