Learn More
Biomatik Corporation Mouse Glucagon Like Peptide 1 (GLP1) ELISA Kit
Supplier:  Biomatik Corporation EKU12153
Encompass_Preferred
Mouse Glucagon Like Peptide 1 (GLP1) ELISA Kit for detecting GLP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.07pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P55095. Gene ID: 14526. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4588