Learn More
Biomatik Corporation Mouse Glucocorticoid Receptor Beta (GRb) ELISA Kit
Supplier:  Biomatik Corporation EKU04399
Encompass_Preferred
Mouse Glucocorticoid Receptor Beta (GRb) ELISA Kit for detecting GRb in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06537. Gene ID: 14815. Target Synonyms: GR-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5137