Learn More
Cayman Chemical Mouse GlucocortIcoId RECPTR
SDP
Catalog No. 502994085
Encompass_Preferred
mGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog No. M00201, M00201-32]
Catalog No. 50-299-4085 Supplier Cayman Chemical Supplier No. 214003X32 WELL
mGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog No. M00201, M00201-32]