Learn More
Cayman Chemical Mouse GlucocortIcoId RECPTR 96
SDP
Catalog No. 502964378
Encompass_Preferred
mGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog No. M00201, M00201-32]
Catalog No. 50-296-4378 Supplier Cayman Chemical Supplier No. 2140096 WELL
Add to Cart
Add to Cart
mGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog No. M00201, M00201-32]