Learn More
American Research Products Inc Mouse glucoprotein 130 gp130
Catalog No. 5000328766
Encompass
Mouse glucoprotein 130 gp130 ELISA KIT from CUSABIO Detection Range 78 pg/mL - 5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method
Catalog No. 50-003-28766 Supplier American Research Products Inc Supplier No. CSBE04572M
Mouse glucoprotein 130 gp130 ELISA KIT from CUSABIO Detection Range 78 pg/mL - 5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method