Learn More
Crystal Chem Inc Mouse Glucose Assay Kit, 10-Kit Pack, 96-tests per well
SDP
Catalog No. 501947909
Encompass_Preferred
The Mouse Glucose 10-kit pack is an economical option for high-volume users of our Mouse Glucose Assay Kit offered as ten individual kits.Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for mouse samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7909 Supplier Crystal Chem Inc Supplier No. 81694
Add to Cart
Add to Cart
The Mouse Glucose 10-kit pack is an economical option for high-volume users of our Mouse Glucose Assay Kit offered as ten individual kits.Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for mouse samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.