Learn More
Crystal Chem Inc Mouse Glucose Assay Kit, 96-tests
SDP
Catalog No. 501947907
Encompass
The Mouse Glucose Assay is used to quantify levels of glucose in mouse serum, plasma, and urine using only a 2 µL sample volume. Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for mouse samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7907 Supplier Crystal Chem Inc Supplier No. 81692
The Mouse Glucose Assay is used to quantify levels of glucose in mouse serum, plasma, and urine using only a 2 µL sample volume. Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for mouse samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.