Learn More
American Research Products Inc Mouse Glucose dependent insuli
Catalog No. 5000326696
Encompass
Mouse Glucose-dependent insulin-releasing polypeptide GIP ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26696 Supplier American Research Products Inc Supplier No. CSBE08486M
Mouse Glucose-dependent insulin-releasing polypeptide GIP ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich