Learn More
American Research Products Inc Mouse Glucose Regulated Protei
Catalog No. 5000327750
Encompass
Mouse Glucose Regulated Protein 78 (GRP78) Elisa kit from CUSABIO Detection Range 0 45 ng/mL-30 ng/mL Sensitivity 0 11 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27750 Supplier American Research Products Inc Supplier No. CSBE17690M
Mouse Glucose Regulated Protein 78 (GRP78) Elisa kit from CUSABIO Detection Range 0 45 ng/mL-30 ng/mL Sensitivity 0 11 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich