Learn More
Biomatik Corporation Mouse Glucuronidase Beta (GUSb) ELISA Kit
Supplier:  Biomatik Corporation EKU04430
Encompass_Preferred
Mouse Glucuronidase Beta (GUSb) ELISA Kit for detecting GUSb in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 23.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12265. Gene ID: 110006. Target Synonyms: GUS-B; MPS7; Beta-G1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8957