Learn More
American Research Products Inc Mouse Glutamate decarboXylase
Catalog No. 5000327301
Encompass
Mouse Glutamate decarboXylase 1 (GAD1) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27301 Supplier American Research Products Inc Supplier No. CSBEL009159MO
Mouse Glutamate decarboXylase 1 (GAD1) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich