Learn More
American Research Products Inc Mouse Glutamate NMDA receptor
Catalog No. 5000327657
Encompass
Mouse Glutamate [NMDA] receptor subunit epsilon-2 (GRIN2B) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27657 Supplier American Research Products Inc Supplier No. CSBEL009913MO
Mouse Glutamate [NMDA] receptor subunit epsilon-2 (GRIN2B) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich