Learn More
Biomatik Corporation Mouse Glutaminase 2 (GLS2) ELISA Kit
Supplier:  Biomatik Corporation EKU04464
Encompass_Preferred
Mouse Glutaminase 2 (GLS2) ELISA Kit for detecting GLS2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q571F8. Gene ID: 216456. Target Synonyms: GA; GLS; LGA; hLGA; L-glutaminase; L-glutamine amidohydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5158