Learn More
Biomatik Corporation Mouse Glutamine synthetase (GS) CLIA Kit
Supplier:  Biomatik Corporation EKU09495
Encompass_Preferred
Mouse Glutamine synthetase (GS) CLIA Kit for detecting GS in serum, plasma, tissue homogenates, cell lysates, cerebrospinal fluid, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.27-200pg/ml. Sensitivity: 0.12pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P15105. Gene ID: 14645. Target Synonyms: GLNS; GLUL; Glutamate-Ammonia Ligase; Glutamate decarboxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5706