Learn More
American Research Products Inc Mouse GlutaredoXin related pro
Catalog No. 5000327058
Encompass
Mouse GlutaredoXin-related protein 5 mitochondrial (GLRX5) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27058 Supplier American Research Products Inc Supplier No. CSBEL009524MO
Mouse GlutaredoXin-related protein 5 mitochondrial (GLRX5) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich