Learn More
American Research Products Inc Mouse glutathione peroXidase
Catalog No. 5000329519
Encompass
Mouse glutathione peroXidase (GSH-PX) ELISA Kit from CUSABIO Detection Range 31 25 U/mL-2000 U/mL Sensitivity 7 8 U/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29519 Supplier American Research Products Inc Supplier No. CSBE13068M
Mouse glutathione peroXidase (GSH-PX) ELISA Kit from CUSABIO Detection Range 31 25 U/mL-2000 U/mL Sensitivity 7 8 U/mL Sample serum plasma tissue homogenates Method Sandwich