Learn More
Biomatik Corporation Mouse Glutathione S Transferase Mu 1 (GSTM1) ELISA Kit
Supplier:  Biomatik Corporation EKU10873
Encompass_Preferred
Mouse Glutathione S Transferase Mu 1 (GSTM1) ELISA Kit for detecting GSTM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.58ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10649. Gene ID: 14862. Target Synonyms: GST-M1; GST1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GTH4; GTM1; H-B; MU1; GST HB subunit 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8275