Learn More
Biomatik Corporation Mouse Glycogen Synthase Kinase 3 Beta (GSK3b) ELISA Kit
Supplier:  Biomatik Corporation EKU11739
Encompass_Preferred
Mouse Glycogen Synthase Kinase 3 Beta (GSK3b) ELISA Kit for detecting GSK3b in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WV60. Gene ID: 56637. Target Synonyms: Serine/threonine-protein kinase GSK3B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8089