Learn More
Biomatik Corporation Mouse Glycosylated Lysosomal Membrane Protein (GLMP) ELISA Kit
Supplier:  Biomatik Corporation EKU03184
Encompass_Preferred
Mouse Glycosylated Lysosomal Membrane Protein (GLMP) ELISA Kit for detecting GLMP in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JHJ3. Gene ID: 56700. Target Synonyms: C1orf85; NCU-G1; Lysosomal protein NCU-G1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6210