Learn More
Cusabio Technology LLC Mouse Glypican-3 GPC-3ELISA
Catalog No. 5000051920
Encompass_Preferred
The Mouse Glypican-3 (GPC-3) ELISA Kit is designed to quantitatively measure GPC3 levels in serum plasma and cell culture supernatants using a sandwich assay format With a detection range of 3 12 ng/mL to 200 ng/mL and a sensitivity of 0 78 ng/mL this kit provides reliable results within 1-5 hours of processing time The assay requires 50-100 L of sample per test and utilizes absorbance measurement at 450 nm for accurate detection Ideal for both research and diagnostic applications it enables precise quantification of GPC3 protein concentrations in various biological matrices while maintaining consistency and reproducibility across experimental conditions
Catalog No. 50-000-51920 Supplier Cusabio Technology LLC Supplier No. CSBE15877M
The Mouse Glypican-3 (GPC-3) ELISA Kit is designed to quantitatively measure GPC3 levels in serum plasma and cell culture supernatants using a sandwich assay format With a detection range of 3 12 ng/mL to 200 ng/mL and a sensitivity of 0 78 ng/mL this kit provides reliable results within 1-5 hours of processing time The assay requires 50-100 L of sample per test and utilizes absorbance measurement at 450 nm for accurate detection Ideal for both research and diagnostic applications it enables precise quantification of GPC3 protein concentrations in various biological matrices while maintaining consistency and reproducibility across experimental conditions