Learn More
ACROBiosystems ActiveMax Mouse GM-CSF Protein, Tag Free
Catalog No. 502331642
Encompass_Preferred
ActiveMax Mouse GM-CSF, Tag Free (GMF-M5211) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Lys 141 (Accession # P01587).
Catalog No. 50-233-1642 Supplier ACROBiosystems Supplier No. GMFM521150UG
ActiveMax Mouse GM-CSF, Tag Free (GMF-M5211) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Lys 141 (Accession # P01587).