Learn More
Kingfisher Biotech Inc Chicken CCL20 ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1498M004
Encompass_Preferred
The Chicken CCL20 (LARC, MIP-3 alpha, Exodus-1) ELISA contains capture antibody, standard, and detection antibody for development of a Chicken CCL20 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include CCL20, CKb4, Exodus, LARC, MIP-3-alpha, MIP-3a, MIP3A, SCYA20, ST38, chemokine (C-C motif) ligand 20, C-C motif chemokine ligand 20. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5500