Learn More
Sino Biological Recombinant Mouse GM-CSF/CSF2 Protein (His Tag) 500
Supplier:  Sino Biological 51048M07H500
Encompass_Preferred
A DNA sequence encoding the mouse CSF2 (P01587) (Ala18-Lys141) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-7189