Learn More
BIOHIPPO Mouse GMCSF Microsample
SDP
Catalog No. 5000313693
Encompass
Mouse GMCSF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GMCSF in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 0 57 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GMCSF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GMCSF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GMCSF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13693 Supplier BIOHIPPO Supplier No. BHE15212336
Mouse GMCSF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GMCSF in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 0 57 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GMCSF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GMCSF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GMCSF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color