Learn More
Raybiotech Inc Mouse gp130 ELISA Kit
Supplier:  Raybiotech Inc ELMGP1301
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 40 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human mouse gp-130. Other species not determined yet
Catalog No. 50-195-0001