Learn More
Sino Biological Mouse GPC3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50989T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Glypican 3 / GPC3 / OCI-5 (rM Glypican 3 / GPC3 / OCI-5; Catalog#50989-M08B; NP_057906.2; Met1-Pro558). Glypican 3 / GPC3 / OCI-5 specific IgG was purified by Mouse Glypican 3 / GPC3 / OCI-5 affinity chromatography.
Catalog No. 50-237-3413