Learn More
BIOHIPPO Mouse GPX7 Microsample
SDP
Catalog No. 5000314801
Encompass
Mouse GPX7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GPX7 in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 07 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GPX7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GPX7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GPX7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14801 Supplier BIOHIPPO Supplier No. BHE15212381
Mouse GPX7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GPX7 in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 07 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GPX7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GPX7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GPX7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color